Learn More
World Precision Instrument WPI Premium Vannas Spring Scissors, 11 cm, Extra Thin 13 mm Blades, Titanium
SDP
Catalog No. NC1656257
Encompass
Our Vannas scissors are specialized spring scissors designed for precision cutting and often used by scientific researchers. They are named after the Swedish scientist Gunnar Vannas who invented them in the 1940s. Compared with other spring scissors, Vannas scissors are distinguished by their delicate, thin, sharp tips. These surgical scissors are 11cm long and have 13mm straight, extra thin blades and sharp tips. The slender, pointed tips allows for precise cutting with minimal damage to surrounding tissues. Choose between straight or angled on the flat blades. Manufactured with surgical grade stainless steel, these scissors are durable and resistant to corrosion. Like all spring scissors, these are an excellent choice for left-handed operators, because they can be used ambidextrously. To properly care for your surgical instruments, these scissors should be cleaned immediately after use to ensure a long service life and minimize staining or corrosion.
This item is not returnable. View return policy
Catalog No. NC1656257 Supplier World Precision Instrument Supplier No. WP5120
Our Vannas scissors are specialized spring scissors designed for precision cutting and often used by scientific researchers. They are named after the Swedish scientist Gunnar Vannas who invented them in the 1940s. Compared with other spring scissors, Vannas scissors are distinguished by their delicate, thin, sharp tips. These surgical scissors are 11cm long and have 13mm straight, extra thin blades and sharp tips. The slender, pointed tips allows for precise cutting with minimal damage to surrounding tissues. Choose between straight or angled on the flat blades. Manufactured with surgical grade stainless steel, these scissors are durable and resistant to corrosion. Like all spring scissors, these are an excellent choice for left-handed operators, because they can be used ambidextrously. To properly care for your surgical instruments, these scissors should be cleaned immediately after use to ensure a long service life and minimize staining or corrosion.