Learn More
Med Vet International Miltex Wagner Plastic Surgery Scissors, Curved, Serrated Blade, Sharp/Blunt, 4.75in, 5-278
Catalog No. 502832486
Encompass_Preferred
Designed specifically for cosmetic and plastic surgery. They are very delicate and lightweight making them ideal for dissecting, cutting, holding and suturing tissue during restorative surgery. Black rubber grips on the rings provide a secure grip. Curved blades with sharp/blunt tips. A serrated finish on the blades help prevent slips. 4.75in total length. Made with high-quality, medical-grade German steel.
Catalog No. 50-283-2486 Supplier Med Vet International Supplier No. MIL5278
Designed specifically for cosmetic and plastic surgery. They are very delicate and lightweight making them ideal for dissecting, cutting, holding and suturing tissue during restorative surgery. Black rubber grips on the rings provide a secure grip. Curved blades with sharp/blunt tips. A serrated finish on the blades help prevent slips. 4.75in total length. Made with high-quality, medical-grade German steel.