Learn More
Med Vet International Miltex Wagner Plastic Surgery Scissors, Straight, Serrated Blade, Sharp/Blunt, 4.75in, 5-273
Catalog No. 502830952
Encompass_Preferred
Designed specifically for cosmetic and plastic surgery. They are very delicate and lightweight making them ideal for dissecting, cutting, holding and suturing tissue during restorative surgery. Black rubber grips on the rings provide a secure grip. Straight blades with sharp/blunt tips. A serrated finish on the blades help prevent slips. 4.75in total length. Made with high-quality, medical-grade German steel.
Catalog No. 50-283-0952 Supplier Med Vet International Supplier No. MIL5273
Designed specifically for cosmetic and plastic surgery. They are very delicate and lightweight making them ideal for dissecting, cutting, holding and suturing tissue during restorative surgery. Black rubber grips on the rings provide a secure grip. Straight blades with sharp/blunt tips. A serrated finish on the blades help prevent slips. 4.75in total length. Made with high-quality, medical-grade German steel.