Learn More
Med Vet International Miltex Westcott Tenotomy Scissors, 5.25in., Right
Catalog No. 502829143
Encompass_Preferred
The Miltex 18-1487 Westcott Tenotomy Scissors are made of German grade stainless steel and measure 5.25in. in length. Westcott Tenotomy Scissors a re a versatile spring-handle thumb scissor that were designed for cutting delicate tissues. Theses scissors have a right angled with blunt/blunt tips
Catalog No. 50-282-9143 Supplier Med Vet International Supplier No. MIL181487
The Miltex 18-1487 Westcott Tenotomy Scissors are made of German grade stainless steel and measure 5.25in. in length. Westcott Tenotomy Scissors a re a versatile spring-handle thumb scissor that were designed for cutting delicate tissues. Theses scissors have a right angled with blunt/blunt tips