Learn More
Med Vet International Miltex Wire Cutting Scissors, Angled To Side, Serrated, German, 4.75in, 9-124
Catalog No. 502831504
Encompass_Preferred
Strong and study to make cutting through suture wires a breeze. Blades are angled to the side and serrated to prevent slippage. 4.75in total length. Made with high-end, medical-grade German steel.
Catalog No. 50-283-1504 Supplier Med Vet International Supplier No. MIL9124
Strong and study to make cutting through suture wires a breeze. Blades are angled to the side and serrated to prevent slippage. 4.75in total length. Made with high-end, medical-grade German steel.