Learn More
Med Vet International 4.25-in Curved Ribbon Iris Scissors, German
Catalog No. 502092740
Encompass_Preferred
Ribbon Iris Scissors were originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal.The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2740 Supplier Med Vet International Supplier No. VK0140110
Ribbon Iris Scissors were originally designed for ophthalmic procedures, Iris Scissors have become a multipurpose instrument used in various surgical environments. These fine, sharp scissors are ideal for detailed dissection of fine tissue and fine suture removal.The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.