Learn More
Med Vet International 6.75-in Economy Mayo Scissors, Straight
Catalog No. 502223960
Encompass_Preferred
Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade.
Catalog No. 50-222-3960 Supplier Med Vet International Supplier No. 4302
Mayo Dissecting Scissors are a versatile instrument that can be used in multiple procedures and settings. They are available straight or curved, in a variety of lengths, and have a standard beveled blade.