Learn More
Med Vet International 5 3/4" Economy Metzenbaum Scissors, Curved
Catalog No. 502224040
Encompass_Preferred
These scissors are designed for cutting delicate tissue and blunt dissection. The scissors have a relatively long shank-to-blade ratio.
Catalog No. 50-222-4040 Supplier Med Vet International Supplier No. 4321
These scissors are designed for cutting delicate tissue and blunt dissection. The scissors have a relatively long shank-to-blade ratio.