Learn More
Med Vet International Or Scissors 5.75in, Curved/ Blunt/Blunt, Von Klaus German Surgical Steel
Catalog No. 502092750
Encompass_Preferred
These surgical scissors are curved and have a blunt/blunt tip and are used generally for cutting sutures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.”
Catalog No. 50-209-2750 Supplier Med Vet International Supplier No. VK1030314
These surgical scissors are curved and have a blunt/blunt tip and are used generally for cutting sutures. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.”