Learn More
Med Vet International Spencer Stitch Scissors, 3.5in, Straight, Delicate Pattern, German Grade, Von Klaus
Catalog No. 502092733
Encompass_Preferred
3.5in. Spencer Stitch Scissors are straight and have a delicate pattern. Spencer Stitch scissors is a German grade surgical instrument and are normally used for suture removal. They feature a small hook to lightly lift up sutures before removing them. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.
Catalog No. 50-209-2733 Supplier Med Vet International Supplier No. VK1140609
3.5in. Spencer Stitch Scissors are straight and have a delicate pattern. Spencer Stitch scissors is a German grade surgical instrument and are normally used for suture removal. They feature a small hook to lightly lift up sutures before removing them. The Von Klaus line of instruments is made of high quality German stainless steel, expertly crafted and comes with a lifetime warranty when used for their intended surgical purpose.