Learn More
PARKER HANNIFIN CORP SCITEMP TEMP SNSR 3/4IN TC 5PK
SDP
Catalog No. NC9950068
Encompass
  • Single-use design for enhanced safety in sensitive environments.
  • Pre-calibrated sensor ensures consistent performance and reliability.
  • Temperature range of 4 to 70 °C for versatile applications.
  • Temperature accuracy of ± 0.5 °C guarantees precise measurements.
  • Built with medical-grade, animal-free compounds for biocompatibility.
  • Specifications meet USP Class VI and FDA 21 CFR 177.1520 standards.
  • Construction from medical grade polysulfone for durability.
  • Utilizes a gamma-stable chip for storing calibration and identity data.
  • Capable of withstanding maximum pressure of 60 psi.
  • Maximum flow rate of 31 lpm accommodates various fluid dynamics.
  • Connection type: 3/4 in Tri-Clover (TC) for easy integration.
Catalog No. NC9950068 Supplier PARKER HANNIFIN CORP Supplier No. 080796PSX5
  • Single-use design for enhanced safety in sensitive environments.
  • Pre-calibrated sensor ensures consistent performance and reliability.
  • Temperature range of 4 to 70 °C for versatile applications.
  • Temperature accuracy of ± 0.5 °C guarantees precise measurements.
  • Built with medical-grade, animal-free compounds for biocompatibility.
  • Specifications meet USP Class VI and FDA 21 CFR 177.1520 standards.
  • Construction from medical grade polysulfone for durability.
  • Utilizes a gamma-stable chip for storing calibration and identity data.
  • Capable of withstanding maximum pressure of 60 psi.
  • Maximum flow rate of 31 lpm accommodates various fluid dynamics.
  • Connection type: 3/4 in Tri-Clover (TC) for easy integration.