Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sclareol glycol | 55881-96-4 | 10 MG
SDP

Catalog No. 5000419160
Encompass_Preferred

Sclareol glycol is a chemical compound that serves as the precursor of ambroxide. It is notably produced by the microorganism Hyphozyma roseonigra ATCC 20624, which is the only reported strain capable of degrading sclareol to yield sclareol glycol.

  • Purity of 98.0%
  • Molecular formula C16H30O2
  • Molecular weight of 254.41
  • Appears as a white to off-white solid
  • Structure consistent with 1H NMR spectrum and MS
  • In vitro solubility in DMSO is 16.67 mg/mL (65.52 mM)

Catalog No. 50-004-19160 Supplier Medchemexpress LLC Supplier No. HYN700710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sclareol glycol is a chemical compound that serves as the precursor of ambroxide. It is notably produced by the microorganism Hyphozyma roseonigra ATCC 20624, which is the only reported strain capable of degrading sclareol to yield sclareol glycol.

  • Purity of 98.0%
  • Molecular formula C16H30O2
  • Molecular weight of 254.41
  • Appears as a white to off-white solid
  • Structure consistent with 1H NMR spectrum and MS
  • In vitro solubility in DMSO is 16.67 mg/mL (65.52 mM)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.