Learn More
Medchemexpress LLC HY-N0129 100mg Medchemexpress, Sclareolide CAS:564-20-5 Purity:>98%
SDP
Catalog No. 501970227
Encompass_Preferred
Medchemexpress, HY-N0129 100mg Sclareolide CAS:564-20-5 Sclareolide is isolated from the flower of Salvia sclarea with antibacterial and cytotoxic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0227 Supplier Medchemexpress LLC Supplier No. HYN0129100MG
Medchemexpress, HY-N0129 100mg Sclareolide CAS:564-20-5 Sclareolide is isolated from the flower of Salvia sclarea with antibacterial and cytotoxic activities. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.