Learn More
Alpco Ltd Sclerostin ELISA, 96 Wells, 20 L

Supplier: Alpco Ltd 04BI20492
The Sclerostin ELISA is intended for the quantitative determination of sclerostin in Human serum, EDTA plasma or heparin plasma.
- Includes controls and standards
- Small sample volume requirement: 20 µL
- Colorimetric Detection
- Excellent precision and reproducibility
- Intra-assay CV 7%
- Inter-assay CV 10%
For Research Use Only. Not for Use in Diagnostic Procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.