Learn More
Enzo Life Sciences LEADING LIGHT Sclerostin-LRP Interaction Screening System (1x96wells)
SDP
Catalog No. 502012656
Encompass_Preferred
The LEADING LIGHT Sclerostin Screening System is a 1x96 well plate assay based on Sclerostin–LRP interaction. This system contains engineered LRP5-alkaline phosphatase (ALP) and human Sclerostin immobilized in a multi-well plate. LRP5-alkaline phosphatase binds to Sclerostin coated on the bottom of the well and is detected by the activity of alkaline phosphatase remaining after a washing step. This system can be used for drug screening to identify small molecule compounds, antibodies, DNA aptamers and peptides capable of modulating sclerostin-LRP interaction. Alternative name: Sclerostin-LRP binding kit, SOST. Assay Time: 2.5 hours. Applications: FUNC, Chemiluminescence, HTS . Species reactivity: Human .
Catalog No. 50-201-2656 Supplier Enzo Life Sciences Supplier No. ENZ610030001
The LEADING LIGHT Sclerostin Screening System is a 1x96 well plate assay based on Sclerostin–LRP interaction. This system contains engineered LRP5-alkaline phosphatase (ALP) and human Sclerostin immobilized in a multi-well plate. LRP5-alkaline phosphatase binds to Sclerostin coated on the bottom of the well and is detected by the activity of alkaline phosphatase remaining after a washing step. This system can be used for drug screening to identify small molecule compounds, antibodies, DNA aptamers and peptides capable of modulating sclerostin-LRP interaction. Alternative name: Sclerostin-LRP binding kit, SOST. Assay Time: 2.5 hours. Applications: FUNC, Chemiluminescence, HTS . Species reactivity: Human .