Learn More
AdipoGen Sclerotiorin (0.5 mg)
SDP
Supplier:  AdipoGen AGCN20054MC05
Encompass_Preferred
Chemical. CAS: 549-23-5. Formula: C21H23ClO5. MW: 390.9. Isolated from fungus Penicillium sp. Antibiotic. Antibacterial. Weak antagonist of human endothelin receptor A (ETA) and B (ETB). Cholesteryl ester transfer protein (CETP) activity inhibitor. Grb2-Shc binding inhibitor. Potent aldose reductase inhibitor. Potent reversible, non-competitive soybean lipoxygenase-1 (LOX-1) inhibitor. Antioxidant. Free radical scavenger. Antifungal.
Catalog No. 50-205-3399