Learn More
ABclonal Technology SCLY Rabbit pAb
SDP
Catalog No. 5000106643
Encompass_Preferred
Selenocysteine lyase (SCLY, EC 4.4.1.16) catalyzes the pyridoxal 5-prime phosphate-dependent conversion of L-selenocysteine to L-alanine and elemental selenium (Mihara et al., 2000 [PubMed 10692412]).
Catalog No. 50-001-06643 Supplier ABclonal Technology Supplier No. A15842500UL
Selenocysteine lyase (SCLY, EC 4.4.1.16) catalyzes the pyridoxal 5-prime phosphate-dependent conversion of L-selenocysteine to L-alanine and elemental selenium (Mihara et al., 2000 [PubMed 10692412]).