Learn More
Aviva Systems Biology Corp SCML4 Antibody - N-terminal region (ARP34497_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34497_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCML4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SCML4. Alternate names: FLJ36252, dJ47M23.1ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55420