Learn More
Aviva Systems Biology Corp SCN1B Antibody - middle region (ARP35028_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35028_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCN1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SCN1B. Alternate names: GEFSP1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-52985