Learn More
Aviva Systems Biology Corp Scn1b Antibody - N-terminal region (ARP35577_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35577_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Scn1b. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N-terminal region of Rat Scn1b. Alternate names: -ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-284-466