Learn More
Aviva Systems Biology Corp SCN3B Antibody - N-terminal region (ARP37699_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37699_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCN3B. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human SCN3B. Alternate names: HSA243396, SCNB3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53089