Learn More
Aviva Systems Biology Corp SCN8A Antibody - middle region (ARP35227_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP35227_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCN8A. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SCN8A. Alternate names: CerIII, MED, NaCh6, Nav1.6, PN4, CIAT, CERIIIApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55674