Learn More
Aviva Systems Biology Corp SCNN1B Antibody - C-terminal region (ARP34936_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34936_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SCNN1B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human SCNN1B. Alternate names: BESC1, ENaCb, SCNEB, ENaCbetaApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-55611