Learn More
Medchemexpress LLC SCO-NHS carbonate | 2925279-71-4 | 95.0% | 265.26 | C13H15NO5 | 10 MG
SDP
Catalog No. 5000287828
Encompass_Preferred
SCO-NHS carbonate is a reagent used in bio-conjugation workflows to introduce a strained cyclooctyne (SCO) carbonate group via NHS carbonate chemistry. It is intended for research use in strain-promoted alkyne-azide cycloaddition (SPAAC) and related click chemistry applications, and is supplied as a solid with good solubility in DMSO for in vitro use.
  • Click-chemistry reagent suitable for bioconjugation and SPAAC.
  • High purity ensures reproducible conjugation performance.
  • White to off-white solid that is easy to handle and store.
  • Soluble in DMSO at high concentration for in vitro applications.
  • Stable under sealed storage away from moisture and light.
Catalog No. 50-002-87828 Supplier Medchemexpress LLC Supplier No. HY15701210MG
SCO-NHS carbonate is a reagent used in bio-conjugation workflows to introduce a strained cyclooctyne (SCO) carbonate group via NHS carbonate chemistry. It is intended for research use in strain-promoted alkyne-azide cycloaddition (SPAAC) and related click chemistry applications, and is supplied as a solid with good solubility in DMSO for in vitro use.
  • Click-chemistry reagent suitable for bioconjugation and SPAAC.
  • High purity ensures reproducible conjugation performance.
  • White to off-white solid that is easy to handle and store.
  • Soluble in DMSO at high concentration for in vitro applications.
  • Stable under sealed storage away from moisture and light.