Learn More
BURKLE INC PTFE Scoop, 0.34 oz. (10 ml), ØxH 0.98x1.34 in. (25x34 mm), length 6.10 in. (155 mm)
Catalog No. 5000026031
Encompass_Preferred
All parts coming into contact with the sample medium are made of pure white PTFE. Completely inert against all substances (with the exception of elementary fluoride). The rod is stabilized with a steel core which is completely encapsulated in PTFE. The scoop PTFE with 10 ml (0.34 oz.) has a strong rod with a length of 155 mm (6.10 in.).
Catalog No. 50-000-26031 Supplier BURKLE INC Supplier No. 53890010
All parts coming into contact with the sample medium are made of pure white PTFE. Completely inert against all substances (with the exception of elementary fluoride). The rod is stabilized with a steel core which is completely encapsulated in PTFE. The scoop PTFE with 10 ml (0.34 oz.) has a strong rod with a length of 155 mm (6.10 in.).