Learn More
BURKLE INC Scoop angular beaker, PP, 67.63 oz. (2000 ml), TeleScoop
Catalog No. 5000026043
Encompass_Preferred
The beaker is adaptable to different container shapes by adjustable angle setting (up to 90°). It is an attachment for the TeleScoop system and compatible with Burkle telescopic rods – perfect for industrial use and water treatment applications.
Catalog No. 50-000-26043 Supplier BURKLE INC Supplier No. 53540200
The beaker is adaptable to different container shapes by adjustable angle setting (up to 90°). It is an attachment for the TeleScoop system and compatible with Burkle telescopic rods – perfect for industrial use and water treatment applications.