Learn More
BURKLE INC Scoop angular beaker, PP, 20.29 oz. (600 ml), TeleScoop
Catalog No. 5000025864
Encompass_Preferred
The beaker is adaptable to different container shapes by adjustable angle setting (up to 90°). It is an attachment for the TeleScoop system and compatible with Burkle telescopic rods – perfect for industrial use and water treatment applications.
Catalog No. 50-000-25864 Supplier BURKLE INC Supplier No. 53540050
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The beaker is adaptable to different container shapes by adjustable angle setting (up to 90°). It is an attachment for the TeleScoop system and compatible with Burkle telescopic rods – perfect for industrial use and water treatment applications.