Learn More
BURKLE INC Scoop angular beaker, PP, 33.81 oz. (1000 ml), TeleScoop
Catalog No. NC2187702
Encompass_Preferred
The beaker is adaptable to different container shapes by adjustable angle setting (up to 90°). It is an attachment for the TeleScoop system and compatible with Burkle telescopic rods – perfect for industrial use and water treatment applications.
This item is not returnable. View return policy
Catalog No. NC2187702 Supplier BURKLE INC Supplier No. 53540100
The beaker is adaptable to different container shapes by adjustable angle setting (up to 90°). It is an attachment for the TeleScoop system and compatible with Burkle telescopic rods – perfect for industrial use and water treatment applications.