Learn More
Uline Scoop - Stainless Steel, 16 ozScoop powders, dry grains and bulk ingredients.Type 304. Resists rust and corrosion.Polished finish is easy to clean.Use with drums and bulk bags.Dishwasher safe.NSF certified.
Catalog No. NC2241631
Encompass
Scoop - Stainless Steel, 16 ozScoop powders, dry grains and bulk ingredients.Type 304. Resists rust and corrosion.Polished finish is easy to clean.Use with drums and bulk bags.Dishwasher safe.NSF certified.
This item is not returnable. View return policy
Catalog No. NC2241631 Supplier Uline Supplier No. H7428
Scoop - Stainless Steel, 16 ozScoop powders, dry grains and bulk ingredients.Type 304. Resists rust and corrosion.Polished finish is easy to clean.Use with drums and bulk bags.Dishwasher safe.NSF certified.