Learn More
Med Vet International OB/GYN Swab Scopettes Jr. 8 Inch Length NonSterile, 34-7021-12, 100/Box
Catalog No. 503042911
Encompass_Preferred
8-inch length Safe, highly absorbent, uniform and lint free Machine made for uniformity, the 5/8-inch lint free rayon swab head has a 3/16-inch cushioned tip to minimize the possibility of patient tissue trauma Single ended Comfort tip 100 nonsterile Scopettes obstetrical/gynecological applicators in a dispenser box
Catalog No. 50-304-2911 Supplier Med Vet International Supplier No. 35628BX
8-inch length Safe, highly absorbent, uniform and lint free Machine made for uniformity, the 5/8-inch lint free rayon swab head has a 3/16-inch cushioned tip to minimize the possibility of patient tissue trauma Single ended Comfort tip 100 nonsterile Scopettes obstetrical/gynecological applicators in a dispenser box