Learn More
BIRCHWOOD LABORATORIES SCOPETTES LRG TIP 8IN STERILE
Supplier:  BIRCHWOOD LABORATORIES 34702250
Encompass
This item has a minimum qty of 2 per supplier requirements.
Scopettes Jr. 8-inch also available in sterile packaging. The applicators are sterilized by ethylene oxide and packaged in bar-coded chevron-style peel pouches containing six swabs each. Case= 50 x package of 6.
Catalog No. NC9895553