Learn More
Medchemexpress LLC Scopine hydrochloride | 85700-55-6 | 99.69% | 50 MG
SDP
Catalog No. 5000405337
Encompass_Preferred
Scopine hydrochloride is a metabolite of Scopolamine and a brain-targeting compound. It significantly increases the brain exposure of Chlorambucil, thereby enhancing its anti-glioma activity. This compound can be utilized in targeted therapy research for brain gliomas.
  • Metabolite of scopolamine
  • Brain-targeting compound
  • Increases brain exposure of Chlorambucil
  • Enhances anti-glioma activity
  • Useful in targeted therapy research for brain gliomas
Catalog No. 50-004-05337 Supplier Medchemexpress LLC Supplier No. HYB0459A50MG
Scopine hydrochloride is a metabolite of Scopolamine and a brain-targeting compound. It significantly increases the brain exposure of Chlorambucil, thereby enhancing its anti-glioma activity. This compound can be utilized in targeted therapy research for brain gliomas.
  • Metabolite of scopolamine
  • Brain-targeting compound
  • Increases brain exposure of Chlorambucil
  • Enhances anti-glioma activity
  • Useful in targeted therapy research for brain gliomas