Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Scopine hydrochloride | 85700-55-6 | 99.7% | 100 MG
SDP

Catalog No. 5000408211
Encompass_Preferred

Scopine hydrochloride is a metabolite of Scopolamine and a brain-targeting compound. It significantly increases the brain exposure of Chlorambucil and enhances its anti-glioma activity. This compound can be used in targeted therapy research for brain gliomas.

  • Metabolite of scopolamine
  • Brain-targeting compound
  • Increases brain exposure of chlorambucil
  • Enhances anti-glioma activity
  • Suitable for targeted therapy research for brain gliomas
  • Molecular weight: 191.66
  • Formula: C8H14ClNO2
  • Appearance: solid, white to off-white
  • For research use only

Catalog No. 50-004-08211 Supplier Medchemexpress LLC Supplier No. HYB0459A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Scopine hydrochloride is a metabolite of Scopolamine and a brain-targeting compound. It significantly increases the brain exposure of Chlorambucil and enhances its anti-glioma activity. This compound can be used in targeted therapy research for brain gliomas.

  • Metabolite of scopolamine
  • Brain-targeting compound
  • Increases brain exposure of chlorambucil
  • Enhances anti-glioma activity
  • Suitable for targeted therapy research for brain gliomas
  • Molecular weight: 191.66
  • Formula: C8H14ClNO2
  • Appearance: solid, white to off-white
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.