Learn More
Medchemexpress LLC Scopine hydrochloride | 85700-55-6 | 99.7% | 5 MG
SDP
Catalog No. 5000408421
Encompass_Preferred
Scopine hydrochloride, also known by its synonym 6,7-Epoxytropine hydrochloride, is recognized as a metabolite of Scopolamine and functions as a brain-targeting compound. This compound has demonstrated the ability to significantly elevate the brain exposure of Chlorambucil and consequently amplify its anti-glioma activity. These properties make Scopine hydrochloride a candidate for targeted therapy research focused on brain gliomas.
  • It is a metabolite of Scopolamine.
  • It acts as a brain-targeting compound.
  • It significantly increases the brain exposure of Chlorambucil.
  • It enhances the anti-glioma activity of Chlorambucil.
  • It is used in targeted therapy research for brain gliomas.
  • CHLS (Scopine combined with Chlorambucil) exhibits significant anti-glioma activity in C6 cells.
  • Its IC50 & target is the α adrenergic receptor.
Catalog No. 50-004-08421 Supplier Medchemexpress LLC Supplier No. HYB0459A5MG
Scopine hydrochloride, also known by its synonym 6,7-Epoxytropine hydrochloride, is recognized as a metabolite of Scopolamine and functions as a brain-targeting compound. This compound has demonstrated the ability to significantly elevate the brain exposure of Chlorambucil and consequently amplify its anti-glioma activity. These properties make Scopine hydrochloride a candidate for targeted therapy research focused on brain gliomas.
  • It is a metabolite of Scopolamine.
  • It acts as a brain-targeting compound.
  • It significantly increases the brain exposure of Chlorambucil.
  • It enhances the anti-glioma activity of Chlorambucil.
  • It is used in targeted therapy research for brain gliomas.
  • CHLS (Scopine combined with Chlorambucil) exhibits significant anti-glioma activity in C6 cells.
  • Its IC50 & target is the α adrenergic receptor.