Learn More
Chemglass Life Sciences Scoping Marine Style Impeller Blade for 15L Bio Reactors used with 8mm Shaft
SDP
Catalog No. 5000083253
Encompass
Marine style impeller blades are available in two styles, Vortexing and Scoping.The 3 through 7 liter sizes fit the 8mm shaft of the agitator assembly, while the 15 liter size fits the 15mm Shaft size.These blades are used primarily for mammalian cell culture work.Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.
Catalog No. 50-000-83253 Supplier Chemglass Life Sciences Supplier No. CLS138537
Marine style impeller blades are available in two styles, Vortexing and Scoping.The 3 through 7 liter sizes fit the 8mm shaft of the agitator assembly, while the 15 liter size fits the 15mm Shaft size.These blades are used primarily for mammalian cell culture work.Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.