Learn More
Chemglass Life Sciences Scoping Marine Style Impeller Blade for 5L & 7L Bio Reactors
SDP
Catalog No. 5000083191
Encompass
Marine style impeller blades are available in two styles, Vortexing and Scoping. The 3 through 7 liter sizes fit the 8mm shaft of the agitator assembly, while the 15 liter size fits the 15mm Shaft size. CLS-1385-36 and CLS-1385-37 are for use with CLS-1385-35 agitator assembly. These blades are used primarily for mammalian cell culture work. Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
Catalog No. 50-000-83191 Supplier Chemglass Life Sciences Supplier No. CLS138206
Marine style impeller blades are available in two styles, Vortexing and Scoping. The 3 through 7 liter sizes fit the 8mm shaft of the agitator assembly, while the 15 liter size fits the 15mm Shaft size. CLS-1385-36 and CLS-1385-37 are for use with CLS-1385-35 agitator assembly. These blades are used primarily for mammalian cell culture work. Stainless steel components manufactured from 316L with an electropolished surface finish of 20Ra or better.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors. Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.