Learn More
Beckman Coulter Scopio X100 Series Full-Field Digital Cell Morphology Scanner
Shop All Beckman Coulter ProductsDescription
Full-Field Digital Imaging
- Dynamic Scan Image providing enhanced assessment quality and added context for comprehensive analysis.
- A seamless integration of CBC results before and during slide review, alongside Scopio's differential
- No trade-off between field-of-view and resolution, allowing for assessment and analysis of cell morphology at an unmatched scale and depth
AI powered Decision Support
- Adaptive Monolayer Detection algorithm, designed to identify the clinically relevant area and adapt for optimal review.
- White Blood Cells DSS for detection and pre-classification into 14 classes.
- Detection of platelets and pre-estimation from 10 fields of view, including the identification of platelet clumps throughout the scan area and the feathered edge.
Seamless LIS Integration
- Advanced digital platform promotes remote review and consultation throughout the secure hospital network.
- Streamline workflows with an enhanced review process and minimized logistical constraints
- Help specialists collaborate across the secured hospital network with easy remote reviews and consultations
- Customize the analysis process for department-specific needs with features like multiple slide review, customizable category naming, seamless LIS integration, and flexible reporting options
Specifications
Specifications
| For Use With (Equipment) | Beckman Coulter Analyzers |
| Product Line | Scopio X100 |
| Throughput | 15 Slides/hr. |
| Includes | Processing Unit and Full-Field PBS |