Learn More
Siemens Healthineers™ Scopio X100HT Digital Cell Morphology
Catalog No. 23313107 Shop All Siemens Healthineers Products
Change view
Click to view available options
For Use With (Application):
Scopio X100HT
Catalog No. For Use With (Application)
23-313-107 Scopio X100HT
1 options
This item is not returnable. View return policy
Catalog No. 23-313-107 Supplier Siemens Healthineers™ Supplier No. 11641990
Add to Cart
Add to Cart
This item is not returnable. View return policy
Digital Cell Morphology platform for the Scopio X100HT.
Scopio is transforming the way you work with the world's first full-field digital cell morphology platforms. By scanning peripheral blood smear (PBS) samples—from the monolayer to the feathered edge—in 100X resolution, you can digitally pan or zoom to seamlessly go from the full context of a slide, down to an individual cell.
  • Scopio's Full-Field Peripheral Blood Smear Application reduces turnaround time by enabling sharing of digitized slides throughout your secure hospital network
  • With AI-powered decision support, both standardization and efficiency are increased, significantly reducing the complexity associated with traditional morphological analysis
TRUSTED_SUSTAINABILITY
Specifications
Type Microscope
For Use With (Application) Scopio X100HT