Learn More
Selleck Chemical LLC Scopolamine HBr trihydrate
SDP
Catalog No. 5000003657
Encompass_Preferred
Scopolamine HBr is a competitive muscarinic acetylcholine receptor antagonist with an IC50 of 55 3 nM
Catalog No. 50-000-03657 Supplier Selleck Chemical LLC Supplier No. S587325MG
Scopolamine HBr is a competitive muscarinic acetylcholine receptor antagonist with an IC50 of 55 3 nM