Learn More
Apexbio Technology LLC Scopolamine hydrobromide 114-49-8 100mg
SDP
Catalog No. 5000094202
Encompass_Preferred
Scopolamine hydrobromide (114-49-8) is a small-molecule antagonist targeting muscarinic acetylcholine receptors and serotonin 5-HT3 receptors It is designed to reversibly block these receptors thereby modulating cholinergic and serotonergic signaling pathways Scopolamine hydrobromide exerts its biological activity primarily through reversible antagonism of muscarinic receptors and inhibition of serotonin 5-HT3 receptor responses In receptor binding studies Scopolamine hydrobromide demonstrates antagonistic activity with reported IC50 values generally ranging from nanomolar to micromolar concentrations depending on the receptor subtype and experimental conditions Based on these pharmacological properties Scopolamine hydrobromide holds research potential in studies of neurotransmission cholinergic signaling gastrointestinal motility assays cognitive impairment models and emesis reflex investigations
Catalog No. 50-000-94202 Supplier Apexbio Technology LLC Supplier No. N2570100
Scopolamine hydrobromide (114-49-8) is a small-molecule antagonist targeting muscarinic acetylcholine receptors and serotonin 5-HT3 receptors It is designed to reversibly block these receptors thereby modulating cholinergic and serotonergic signaling pathways Scopolamine hydrobromide exerts its biological activity primarily through reversible antagonism of muscarinic receptors and inhibition of serotonin 5-HT3 receptor responses In receptor binding studies Scopolamine hydrobromide demonstrates antagonistic activity with reported IC50 values generally ranging from nanomolar to micromolar concentrations depending on the receptor subtype and experimental conditions Based on these pharmacological properties Scopolamine hydrobromide holds research potential in studies of neurotransmission cholinergic signaling gastrointestinal motility assays cognitive impairment models and emesis reflex investigations