Learn More
Medchemexpress LLC Scopolamine hydrobromide trihydrate | 6533-68-2 | 99.86% | 438.31 | 5 MG
SDP
Catalog No. 5000366336
Encompass_Preferred
Scopolamine (Hyoscine) hydrobromide trihydrate is a high-affinity muscarinic antagonist that can cross the blood-brain barrier. It competitively antagonizes 5-HT3 receptors with an IC50 of 2.09 μM. It can induce cognitive and memory deficits in animals and is used in research for preventing postoperative nausea and vomiting, motion sickness, and nervous system diseases. This product is for research use only and not sold to patients.
  • High-affinity muscarinic antagonist
  • Can cross the blood-brain barrier
  • Competitively antagonizes 5-HT3 receptors
  • Induces cognitive and memory deficits in animals, useful for research in these areas
  • Used in research for preventing postoperative nausea and vomiting, motion sickness, nervous system diseases
Catalog No. 50-003-66336 Supplier Medchemexpress LLC Supplier No. HYW0108925MG
Scopolamine (Hyoscine) hydrobromide trihydrate is a high-affinity muscarinic antagonist that can cross the blood-brain barrier. It competitively antagonizes 5-HT3 receptors with an IC50 of 2.09 μM. It can induce cognitive and memory deficits in animals and is used in research for preventing postoperative nausea and vomiting, motion sickness, and nervous system diseases. This product is for research use only and not sold to patients.
  • High-affinity muscarinic antagonist
  • Can cross the blood-brain barrier
  • Competitively antagonizes 5-HT3 receptors
  • Induces cognitive and memory deficits in animals, useful for research in these areas
  • Used in research for preventing postoperative nausea and vomiting, motion sickness, nervous system diseases