Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(-)-Scopolamine hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111017958 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-7958 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-7958 Supplier MilliporeSigma Supelco Supplier No. 370220.1MG
Add to Cart
Add to Cart

(−)-Scopolamine hydrochloride belongs to the class of naturally occurring tropane alkaloids of medicinal interest, found in several members of the Solanaceae family, commonly Atropa belladonna and representatives of the genera Datura and Duboisia.

Featured Industries: Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 55-16-3
Physical Form Neat
Linear Formula C17H21 NO4 · HCl
MDL Number MFCD00058096
Synonym Hyoscine hydrochloride; Scopine tropate
Type Analytical Standard
Quantity 0.1 mg
Formula Weight 339.81
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.