Learn More
Selleck Chemical LLC Scopolamine LSM-4015 HBr-50mg
SDP
Supplier:  Selleck Chemical LLC S250850MG
Encompass_Preferred
Scopolamine HBr (LSM-4015,NSC 61806,(-)-Scopolamine hydrobromide, Hyoscine hydrobromide, Scopine hydrobromide) is a competitive muscarinic acetylcholine receptor antagonist with an IC50 of 55.3 nM.
Catalog No. 50-238-9970