Learn More
Medchemexpress LLC Scopolamine methyl nitrate | 6106-46-3 | MFCD00078239 | 99.8% | 380.39 g·mol⁻¹ | C18H24N2O7 | 25 MG
SDP
Catalog No. 5000283631
Encompass_Preferred
Scopolamine methyl nitrate (methscopolamine nitrate) is a quaternary ammonium antimuscarinic compound used in life-science research. It is employed in neuroscience and pharmacology to probe muscarinic acetylcholine receptor function and as a biochemical reagent in drug-discovery and assay development.
  • Quaternary ammonium antimuscarinic reagent for in vitro studies.
  • High reported purity suitable for analytical and biochemical applications.
  • Molecular weight 380.39 g·mol⁻¹; formula C18H24N2O7.
  • Available in small research pack sizes (25 mg) for screening and assays.
  • Commonly used to study acetylcholine receptor pharmacology and signaling.
Catalog No. 50-002-83631 Supplier Medchemexpress LLC Supplier No. HYW12770225MG
Scopolamine methyl nitrate (methscopolamine nitrate) is a quaternary ammonium antimuscarinic compound used in life-science research. It is employed in neuroscience and pharmacology to probe muscarinic acetylcholine receptor function and as a biochemical reagent in drug-discovery and assay development.
  • Quaternary ammonium antimuscarinic reagent for in vitro studies.
  • High reported purity suitable for analytical and biochemical applications.
  • Molecular weight 380.39 g·mol⁻¹; formula C18H24N2O7.
  • Available in small research pack sizes (25 mg) for screening and assays.
  • Commonly used to study acetylcholine receptor pharmacology and signaling.