Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scopolamine N-Oxide Hydrobromide Monohydrate 98.0+%, TCI America™
SDP

Catalog No. S02291GTCI Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S02291GTCI 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S02291GTCI Supplier TCI America Supplier No. S02291G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6106-81-6
Molecular Formula C17H22BrNO5
Molecular Weight (g/mol) 400.27
MDL Number MFCD00050590
InChI Key MGNNYKWRWHQLCR-XWDDIHSFSA-N
Synonym Genoscopolamine Hydrobromide, Scopolamine Aminoxide Hydrobromide
PubChem CID 91659059
IUPAC Name hydrogen (1S,2R,4S,5S)-7-{[(2S)-3-hydroxy-2-phenylpropanoyl]oxy}-9-methyl-3-oxa-9-azatricyclo[3.3.1.0²,⁴]nonan-9-ium-9-olate bromide
SMILES [H+].[Br-].C[N+]1([O-])[C@H]2CC(C[C@H]1[C@@H]1O[C@H]21)OC(=O)[C@H](CO)C1=CC=CC=C1

Specifications

Melting Point 137°C
Color White
Quantity 1 g
UN Number 1544
Formula Weight 400.27
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Scopolamine N-Oxide Hydrobromide Monohydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.