Learn More
Medchemexpress LLC HY-N0342 100mg Medchemexpress, Scopoletin CAS:92-61-5 Purity:>98%
SDP
Catalog No. 501933529
Encompass_Preferred
Medchemexpress, HY-N0342 100mg Scopoletin CAS:92-61-5 Scopoletin is an inhibitor of acetylcholinesterase ( AChE ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3529 Supplier Medchemexpress LLC Supplier No. HYN0342100MG
Medchemexpress, HY-N0342 100mg Scopoletin CAS:92-61-5 Scopoletin is an inhibitor of acetylcholinesterase ( AChE ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.