Learn More
TARGETMOL CHEMICALS INC Scopoletin 200MG
SDP
Catalog No. 502876549
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Scopoletin (Esculetin 6-methyl ether) is a plant growth factor derived from the root of Scopolia carniolica, inhibits of acetylcholinesterase (AChE). Purity 99.88%
Catalog No. 50-287-6549 Supplier TARGETMOL CHEMICALS INC Supplier No. T2984200MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Scopoletin (Esculetin 6-methyl ether) is a plant growth factor derived from the root of Scopolia carniolica, inhibits of acetylcholinesterase (AChE). Purity 99.88%