Learn More
Medchemexpress LLC Scopoletin | 92-61-5 | MFCD00006872 | 99.9% | 192.17 | 200 MG
SDP
Catalog No. 5000427211
Encompass_Preferred
Scopoletin is an inhibitor of acetylcholinesterase (AChE). This compound is used in research to study its effects on cellular processes, including proliferation and apoptosis in various cell lines, and its impact on cognitive functions in animal models.
  • Enhances K+-stimulated release of ACh from rat frontal cortex synaptosomes.
  • Inhibits PC3 proliferation by inducing apoptosis of PC3 cells.
  • Increases T-maze alternation and ameliorates scopolamine-induced cholinergic deficit and age-associated deficits in object memory in mice.
  • Available as solid or solution formulations in various quantities.
  • For research use only.
Catalog No. 50-004-27211 Supplier Medchemexpress LLC Supplier No. HYN0342200MG
Scopoletin is an inhibitor of acetylcholinesterase (AChE). This compound is used in research to study its effects on cellular processes, including proliferation and apoptosis in various cell lines, and its impact on cognitive functions in animal models.
  • Enhances K+-stimulated release of ACh from rat frontal cortex synaptosomes.
  • Inhibits PC3 proliferation by inducing apoptosis of PC3 cells.
  • Increases T-maze alternation and ameliorates scopolamine-induced cholinergic deficit and age-associated deficits in object memory in mice.
  • Available as solid or solution formulations in various quantities.
  • For research use only.