Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Scopoletin, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111015913 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-5913 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-5913 Supplier MilliporeSigma Supelco Supplier No. 3833210MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Scopoletin is a coumarin compound and pharmacologically active chemical that is found in various plant species.

Food and Beverages
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 92-61-5
Physical Form Neat
Linear Formula C10H8O4
MDL Number MFCD00006872
Melting Point 203°C to 205°C
Synonym 6-Methoxyumbelliferone; 6-Methylesculetin; 7-Hydroxy-6-methoxy-2 H-1-benzopyran-2-one; 7-Hydroxy-6-methoxycoumarin; Buxuletin; Chrysatropic acid; Escopoletin; Esculetin-6-methyl ether; Gelseminic acid; Murrayetin
Type Analytical Standard
Quantity 10 mg
Formula Weight 192.17
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.