Learn More
Cayman Chemical Scopulaops F244 ymglusn 2.5mg
SDP
Catalog No. 502952059
Encompass_Preferred
A fungal β-lactone antibiotic that inhibits HMG-CoA synthase (IC50 = 0.12 μM) by covalently modifying the active Cys129 residue of the enzyme; targets cell walls of pathogens such as S. aureus in a mouse sepsis model of infection
Catalog No. 50-295-2059 Supplier Cayman Chemical Supplier No. 118992.5 MG
A fungal β-lactone antibiotic that inhibits HMG-CoA synthase (IC50 = 0.12 μM) by covalently modifying the active Cys129 residue of the enzyme; targets cell walls of pathogens such as S. aureus in a mouse sepsis model of infection